accessibility menu, dialog, popup

UserName

H3COONHN

CAS RN 4498-68-4

IUPAC Name: ethyl 1H-indazole-3-carboxylate
Synonyms: ethyl 1H-indazole-3-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: YLKPTYMNELPKOL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results