CAS RN 4483-47-0
CAS RN 4483-47-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diisohomoeugenol, TRC
CAS: 4483-47-0 Molecular Formula: C22H28O4 Molecular Weight (g/mol): 356.46 Synonym: Dimethyl Ether of Diisoeugenol,NSC 10200,1-(3,4-Dimethoxyphenyl)-3-ethyl-2,3-dihydro-5,6-dimethoxy-2-methyl-1H-indene,3-Ethyl-5,6-dimethoxy-1- (3,4-dimethoxyphenyl-2 methyl) -indane SMILES: CCC1C(C)C(c2ccc(OC)c(OC)c2)c3cc(OC)c(OC)cc13