accessibility menu, dialog, popup

UserName

H3CMgCl

CAS RN 44767-62-6

IUPAC Name: magnesium(2+) hexan-1-ide chloride
Synonyms: magnesium(2+) hexan-1-ide chloride
Molecular Weight (g/mol): 144.93
Molecular Formula: C6H13ClMg
InChi Key: GBRJQTLHXWRDOV-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CCCCC[CH2-]
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results