accessibility menu, dialog, popup

UserName

CAS RN 4461-33-0

OCNO

CAS RN 4461-33-0

IUPAC Name: benzoyl isocyanate
Synonyms: benzoyl isocyanate
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: LURYMYITPCOQAU-UHFFFAOYSA-N
SMILES: O=C=NC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)