accessibility menu, dialog, popup

UserName

ClONCO

CAS RN 4461-30-7

IUPAC Name: 2-chloroethanecarbonyl isocyanate
Synonyms: chloroacetyl isocyanate
Molecular Weight (g/mol): 119.5
Molecular Formula: C3H2ClNO2
InChi Key: MOVMEFHWBOWMFU-UHFFFAOYSA-N
SMILES: ClCC(=O)N=C=O
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results