accessibility menu, dialog, popup

UserName

CAS RN 4441-12-7

OPNONONO

CAS RN 4441-12-7

IUPAC Name: 4-[bis(morpholin-4-yl)phosphoroso]morpholine
Synonyms: 4-[bis(morpholin-4-yl)phosphoroso]morpholine
Molecular Weight (g/mol): 305.32
Molecular Formula: C12H24N3O4P
InChi Key: WXMQHPKQCPCDQO-UHFFFAOYSA-N
SMILES: O=P(N1CCOCC1)(N1CCOCC1)N1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)