CAS RN 4425-82-5
CAS RN 4425-82-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-Methylidenefluorene, TRC
CAS: 4425-82-5 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 Synonym: 1,2:3,4-Dibenzofulvene,9-Methylene-9H-fluorene,9-Methylenefluorene,Dibenzfulvene,Dibenzofulvene,Fluorenylidenemethane IUPAC Name: 9-methylidenefluorene SMILES: C=C1C2=C(C=CC=C2)C3=C1C=CC=C3