accessibility menu, dialog, popup

UserName

CAS RN 4412-91-3

HOO

CAS RN 4412-91-3

IUPAC Name: (furan-3-yl)methanol
Synonyms: 3-hydroxymethylfuran
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: STJIISDMSMJQQK-UHFFFAOYSA-N
SMILES: OCC1=COC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)