accessibility menu, dialog, popup

UserName

CAS RN 4398-65-6

H3CCH3CH3Cl

CAS RN 4398-65-6

IUPAC Name: 2-chloro-2-methylhexane
Synonyms: 2-chloro-2-methylhexane
Molecular Weight (g/mol): 134.65
Molecular Formula: C7H15Cl
InChi Key: KBOBQLJBYKKAPN-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results