accessibility menu, dialog, popup

UserName

CH3H2NOO

CAS RN 4389-50-8

IUPAC Name: 2-amino-6-methylbenzoate
Synonyms: 2-amino-6-methylbenzoate
Molecular Weight (g/mol): 150.16
Molecular Formula: C8H8NO2
InChi Key: XHYVBIXKORFHFM-UHFFFAOYSA-M
SMILES: CC1=CC=CC(N)=C1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results