accessibility menu, dialog, popup

UserName

H3COOHOOH3CHO

CAS RN 4385-56-2

IUPAC Name: 2-(4-hydroxy-3,5-dimethoxyphenyl)acetic acid
Synonyms: (4-hydroxy-3,5-dimethoxyphenyl)acetic acid
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: BQBQKSSTFGCRQL-UHFFFAOYSA-N
SMILES: COC1=CC(CC(O)=O)=CC(OC)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results