accessibility menu, dialog, popup

UserName

H3CNHCH3CH3

CAS RN 4385-04-0

IUPAC Name: hexyldimethylazanium
Synonyms: hexyldimethylazanium
Molecular Weight (g/mol): 130.25
Molecular Formula: C8H20N
InChi Key: QMHNQZGXPNCMCO-UHFFFAOYSA-O
SMILES: CCCCCC[NH+](C)C
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results