accessibility menu, dialog, popup

UserName

H3COH

CAS RN 4352-42-5

IUPAC Name: 1-cyclohexylbutan-1-ol
Synonyms: 1-cyclohexylbutan-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: MTUCYAOJXPTLHZ-UHFFFAOYNA-N
SMILES: CCCC(O)C1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results