accessibility menu, dialog, popup

UserName

ONHNI

CAS RN 4349-07-9

IUPAC Name: 5-iodo-3,4-dihydropyrimidin-4-one
Synonyms: 5-iodo-3H-pyrimidin-4-one
Molecular Weight (g/mol): 221.99
Molecular Formula: C4H3IN2O
InChi Key: SMIGOGPUTQXOKF-UHFFFAOYSA-N
SMILES: IC1=CN=CNC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results