accessibility menu, dialog, popup

UserName

CAS RN 4341-24-6

CH3(S)OOH

CAS RN 4341-24-6

IUPAC Name: (5S)-3-hydroxy-5-methylcyclohex-2-en-1-one
Synonyms: (5S)-3-hydroxy-5-methylcyclohex-2-en-1-one
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: MNOUUYHZNVGKSQ-YFKPBYRVSA-N
SMILES: C[C@H]1CC(O)=CC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)