accessibility menu, dialog, popup

UserName

CAS RN 433-27-2

H3COOHFFF

CAS RN 433-27-2

IUPAC Name: 1-ethoxy-2,2,2-trifluoroethan-1-ol
Synonyms: 1-ethoxy-2,2,2-trifluoroethanol
Molecular Weight (g/mol): 144.09
Molecular Formula: C4H7F3O2
InChi Key: KLXJPQNHFFMLIG-UHFFFAOYNA-N
SMILES: CCOC(O)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)