accessibility menu, dialog, popup

UserName

CAS RN 4316-42-1

H3CNN

CAS RN 4316-42-1

IUPAC Name: 1-butyl-1H-imidazole
Synonyms: 1-butylimidazole
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: MCMFEZDRQOJKMN-UHFFFAOYSA-N
SMILES: CCCCN1C=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)