accessibility menu, dialog, popup

UserName

CAS RN 4315-07-5

OOOFFF

CAS RN 4315-07-5

IUPAC Name: 2-[4-(trifluoromethoxy)phenyl]acetate
Synonyms: 2-[4-(trifluoromethoxy)phenyl]acetate
Molecular Weight (g/mol): 219.14
Molecular Formula: C9H6F3O3
InChi Key: ZMTIBXQPXBUWPQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=C(OC(F)(F)F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results