accessibility menu, dialog, popup

UserName

ClON

CAS RN 43111-32-6

IUPAC Name: 2-(3-chlorophenoxy)acetonitrile
Synonyms: 2-(3-chlorophenoxy)acetonitrile
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: NHHNUPMHGVIQQC-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(OCC#N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results