accessibility menu, dialog, popup

UserName

CAS RN 4298-08-2

OH(S)HN(S)HOO

CAS RN 4298-08-2

IUPAC Name: (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid
Synonyms: trans-3-hydroxy-L-proline L-threo-3-hydroxyproline
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: BJBUEDPLEOHJGE-IMJSIDKUSA-N
SMILES: O[C@H]1CCN[C@@H]1C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)