CAS RN 42916-66-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(4-Trifluoromethylphenyl)pentan-1-one, TRC
CAS: 42916-66-5 Molecular Formula: C12H13F3O Molecular Weight (g/mol): 230.23 Synonym: 1-[4-(Trifluoromethyl)phenyl]-1-pentanone,p-(Trifluoromethyl)valerophenone,1-(4-Trifluoromethylphenyl)pentan-1-one IUPAC Name: 1-(4-(trifluoromethyl)phenyl)pentan-1-one SMILES: O=C(CCCC)C1=CC=C(C(F)(F)F)C=C1