accessibility menu, dialog, popup

UserName

CH3OOClFFFNOO

CAS RN 42874-03-3

IUPAC Name: 4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-ethoxy-1-nitrobenzene
Synonyms: oxyfluorfen
Molecular Weight (g/mol): 361.7
Molecular Formula: C15H11ClF3NO4
InChi Key: OQMBBFQZGJFLBU-UHFFFAOYSA-N
SMILES: CCOC1=CC(OC2=CC=C(C=C2Cl)C(F)(F)F)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results