accessibility menu, dialog, popup

UserName

CAS RN 42521-09-5

IUPAC Name: methyl 2,6-dichloropyridine-4-carboxylate
Synonyms: methyl 2,6-dichloropyridine-4-carboxylate
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: XSKGHSUHOYEBTK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=NC(Cl)=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results