CAS RN 4249-19-8
CAS RN 4249-19-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(+)-alpha-Methyl-Beta-alanine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-a-Methyl-b-alanine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-b-aminoisobutyric acid, MedChemExpress
MedChemExpress (S)-b-aminoisobutyric acid is a non-protein amino acid originating from the catabolism of thymine and valine.