missing translation for 'onlineSavingsMsg'
Learn More

(S)-b-aminoisobutyric acid, MedChemExpress

Product Code. 16407517
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16407517 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407517 Supplier Medchem Express Supplier No. HY113380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-b-aminoisobutyric acid is a non-protein amino acid originating from the catabolism of thymine and valine.

(S)-b-aminoisobutyric acid is a non-protein amino acid originating from the catabolism of thymine and valine.

Specifications

Chemical Name or Material (S)-b-aminoisobutyric acid
Physical Form Solid
CAS 4249-19-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C4H9NO2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 250 mg/mL (2424.36 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](C)CN
Molecular Weight (g/mol) 103.12
Formula Weight 103.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.