accessibility menu, dialog, popup

UserName

CAS RN 4249-10-9

CH3CH3H3CCH3

CAS RN 4249-10-9

IUPAC Name: 1,2,3,4-tetramethylcyclopenta-1,3-diene
Synonyms: 1,2,3,4-tetramethylcyclopenta-1,3-diene
Molecular Weight (g/mol): 122.21
Molecular Formula: C9H14
InChi Key: VNPQQEYMXYCAEZ-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(C)C1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (3)