accessibility menu, dialog, popup

UserName

CAS RN 4244-84-2

H2NOOCH3HCl

CAS RN 4244-84-2

IUPAC Name: hydrogen ethyl 3-aminopropanoate chloride
Synonyms: hydrogen β-alanine ethyl ester chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: RJCGNNHKSNIUAT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=O)CCN
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)