accessibility menu, dialog, popup

UserName

CAS RN 423768-49-4

CH3NNFOCl

CAS RN 423768-49-4

IUPAC Name: 1-(4-fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Synonyms: 1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride
Molecular Weight (g/mol): 238.65
Molecular Formula: C11H8ClFN2O
InChi Key: INIAJXQDJISNJB-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=C(F)C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results