accessibility menu, dialog, popup

UserName

CH3SOOH3CSOOCl

CAS RN 423768-46-1

IUPAC Name: methyl 5-(chlorosulfonyl)-4-methylthiophene-2-carboxylate
Synonyms: methyl 5-(chlorosulfonyl)-4-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 254.7
Molecular Formula: C7H7ClO4S2
InChi Key: YWBXGHFHMHUSKH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C)=C(S1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results