accessibility menu, dialog, popup

UserName

CAS RN 423768-43-8

IUPAC Name: 2-bromo-1-[4-methyl-2-(pyrazin-2-yl)-1,3-thiazol-5-yl]ethan-1-one
Synonyms: 2-bromo-1-[4-methyl-2-(pyrazin-2-yl)-1,3-thiazol-5-yl]ethanone
Molecular Weight (g/mol): 298.16
Molecular Formula: C10H8BrN3OS
InChi Key: PQWKERRFBZJRSD-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=NC=CN=C1)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results