accessibility menu, dialog, popup

UserName

ClO

CAS RN 42348-86-7

IUPAC Name: 5-chloro-2,3-dihydro-1H-inden-1-one
Synonyms: 5-chloro-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 166.6
Molecular Formula: C9H7ClO
InChi Key: MEDSHTHCZIOVPU-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=O)CCC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)