accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFFFBr

CAS RN 423-55-2

IUPAC Name: 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
Synonyms: perflubron 1-bromoperfluorooctane PFOB LiquiVent perfluorocaprylyl bromide bromure de n-perfluorooctyle imagent GI
Molecular Weight (g/mol): 498.97
Molecular Formula: C8BrF17
InChi Key: WTWWXOGTJWMJHI-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)