accessibility menu, dialog, popup

UserName

CAS RN 42152-46-5

IUPAC Name: copper(2+) benzene ditrifluoromethanesulfonate
Synonyms: copper(2+) benzene ditriflate
Molecular Weight (g/mol): 439.79
Molecular Formula: C8H6CuF6O6S2
InChi Key: DIMCZVTXAXZJCA-UHFFFAOYSA-L
SMILES: [Cu++].C1=CC=CC=C1.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results