accessibility menu, dialog, popup

UserName

CAS RN 42142-52-9

IUPAC Name: [(3R)-3-hydroxy-3-phenylpropyl](methyl)azanium
Synonyms: [(3R)-3-hydroxy-3-phenylpropyl](methyl)azanium
Molecular Weight (g/mol): 166.24
Molecular Formula: C10H16NO
InChi Key: XXSDCGNHLFVSET-SNVBAGLBSA-O
SMILES: C[NH2+]CC[C@@H](O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results