CAS RN 42139-37-7
IUPAC Name:
1-[(1E)-2-nitroethenyl]-4-(propan-2-yl)benzene
Synonyms:
1-isopropyl-4-[(1E)-2-nitroethenyl]benzene
Molecular Weight (g/mol):
191.23
Molecular Formula:
C11H13NO2
InChi Key:
PLOZMGIWCWVROY-BQYQJAHWSA-N
SMILES:
CC(C)C1=CC=C(\C=C\[N+]([O-])=O)C=C1
1
–
1
of
1
results
1
4-Isopropyl-beta-nitrostyrene, 97%, Thermo Scientific Chemicals
CAS: 42139-37-7 Molecular Formula: C11H13NO2 Molecular Weight (g/mol): 191.23 MDL Number: MFCD00085100 InChI Key: PLOZMGIWCWVROY-BQYQJAHWSA-N Synonym: 1-isopropyl-4-2-nitrovinyl benzene,4-isopropyl-w-nitrostyrene,1-isopropyl-4-e-2-nitroethenyl benzene,1-e-2-nitroethenyl-4-propan-2-ylbenzene,xi-4-isopropyl-beta-nitro-styrene,1-isopropyl-4-2-nitroethenyl benzene,1-isopropyl-4-e-2-nitrovinyl benzene,4-isopropyl-omega-nitrostyrene,hg^dahahenr vuvevf@bhafp,4-isopropyl-beta-nitrostyrene PubChem CID: 5702924 IUPAC Name: 1-[(E)-2-nitroethenyl]-4-propan-2-ylbenzene SMILES: CC(C)C1=CC=C(\C=C\[N+]([O-])=O)C=C1