accessibility menu, dialog, popup

UserName

CAS RN 421-50-1

IUPAC Name: 1,1,1-trifluoropropan-2-one
Synonyms: 1,1,1-trifluoroacetone
Molecular Weight (g/mol): 112.05
Molecular Formula: C3H3F3O
InChi Key: FHUDAMLDXFJHJE-UHFFFAOYSA-N
SMILES: CC(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)