accessibility menu, dialog, popup

UserName

H3COO

CAS RN 4208-57-5

IUPAC Name: 1-(furan-2-yl)butan-1-one
Synonyms: 1-(furan-2-yl)butan-1-one
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: GONWJZJNVDRECJ-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results