accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 42079-78-7

IUPAC Name: 5-methoxy-2-phenyl-4H-chromen-4-one
Synonyms: 5-methoxy-2-phenylchromen-4-one
Molecular Weight (g/mol): 252.27
Molecular Formula: C16H12O3
InChi Key: XRQSPUXANRGDAV-UHFFFAOYSA-N
SMILES: COC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results