accessibility menu, dialog, popup

UserName

CAS RN 42036-78-2

BrH3CP

CAS RN 42036-78-2

IUPAC Name: octyltriphenylphosphanium bromide
Synonyms: octyltriphenylphosphanium bromide
Molecular Weight (g/mol): 455.42
Molecular Formula: C26H32BrP
InChi Key: OBLXVLWZBMAMHE-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)