accessibility menu, dialog, popup

UserName

H2NN

CAS RN 420-04-2

IUPAC Name: aminoformonitrile
Synonyms: cyanamide carbimide, calcium calcium salt cyanamide cyanogen nitride epa pesticide chemical code 014002 caswell no. 485A temposil calcium cyanamide citrated TsAKS citrated calcium cyanamide
Molecular Weight (g/mol): 42.04
Molecular Formula: CH2N2
InChi Key: XZMCDFZZKTWFGF-UHFFFAOYSA-N
SMILES: NC#N
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)