CAS RN 41859-58-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Bezafibrate Ethyl Ester, TRC
CAS: 41859-58-9 Molecular Formula: C21 H24 Cl N O4 Molecular Weight (g/mol): 389.87 Synonym: Ethyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate,Bezafibrate Imp. D (EP) IUPAC Name: ethyl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1
2
Bezafibrate EP Impurity D, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.