accessibility menu, dialog, popup

UserName

CAS RN 41806-40-0

CH3NNOO

CAS RN 41806-40-0

IUPAC Name: 1-methyl-1H-imidazole-5-carboxylate
Synonyms: 3-methylimidazole-4-carboxylate
Molecular Weight (g/mol): 125.11
Molecular Formula: C5H5N2O2
InChi Key: PBEDVTDUVXFSMW-UHFFFAOYSA-M
SMILES: CN1C=NC=C1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results