accessibility menu, dialog, popup

UserName

CAS RN 41693-47-4

H3CCH3OCl

CAS RN 41693-47-4

IUPAC Name: 2-methylhexanoyl chloride
Synonyms: 2-methylhexanoyl chloride
Molecular Weight (g/mol): 148.63
Molecular Formula: C7H13ClO
InChi Key: WKEBLSOTVYFYKY-UHFFFAOYNA-N
SMILES: CCCCC(C)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results