accessibility menu, dialog, popup

UserName

CAS RN 41680-34-6

NH2NHNOO

CAS RN 41680-34-6

IUPAC Name: 5-amino-1H-pyrazole-4-carboxylate
Synonyms: 3-amino-2H-pyrazole-4-carboxylate
Molecular Weight (g/mol): 126.1
Molecular Formula: C4H4N3O2
InChi Key: KMRVTZLKQPFHFS-UHFFFAOYSA-M
SMILES: NC1=C(C=NN1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results