accessibility menu, dialog, popup

UserName

CAS RN 4166-53-4

CH3OOO

CAS RN 4166-53-4

IUPAC Name: 4-methyloxane-2,6-dione
Synonyms: 4-methyloxane-2,6-dione
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: MGICRVTUCPFQQZ-UHFFFAOYSA-N
SMILES: CC1CC(=O)OC(=O)C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)