accessibility menu, dialog, popup

UserName

CAS RN 41587-84-2

IUPAC Name: nickel(2+) bis(2-hydroxyacetate)
Synonyms: nickel(2+) diglycolate
Molecular Weight (g/mol): 208.78
Molecular Formula: C4H6NiO6
InChi Key: OZHLXQFAHIYYDJ-UHFFFAOYSA-L
SMILES: [Ni++].OCC([O-])=O.OCC([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results