accessibility menu, dialog, popup

UserName

CAS RN 41513-05-7

IUPAC Name: N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
Synonyms: N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 282.06
Molecular Formula: C9H7BrF3NO
InChi Key: AOHSBSXIGFIMNC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results