CAS RN 4149-06-8
CAS RN 4149-06-8
IUPAC Name:
3-amino-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms:
5-amino-2-phenyl-4H-pyrazol-3-one
Molecular Weight (g/mol):
175.19
Molecular Formula:
C9H9N3O
InChi Key:
LPOVZHYARSAVIZ-UHFFFAOYSA-N
SMILES:
NC1=NN(C(=O)C1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Amino-1-phenyl-2-pyrazolin-5-one, 97%
CAS: 4149-06-8 Molecular Formula: C9H9N3O Molecular Weight (g/mol): 175.19 MDL Number: MFCD00003137 InChI Key: LPOVZHYARSAVIZ-UHFFFAOYSA-N Synonym: 3-amino-1-phenyl-2-pyrazolin-5-one,3-amino-1-phenyl-1h-pyrazol-5 4h-one,3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-phenyl,3-amino-1-phenyl-4,5-dihydro-1h-pyrazol-5-one,5-pyrazolone, 3-amino-1-phenyl,5-amino-2-phenyl-2,4-dihydro-3h-pyrazol-3-one,3h-pyrazol-3-one, 2,4-dihydro-5-amino-2-phenyl,2,4-dihydro-5-amino-2-phenyl-3h-pyrazol-3-one,acmc-1arey,lpovzhyarsaviz-uhfffaoysa PubChem CID: 77794 IUPAC Name: 5-amino-2-phenyl-4H-pyrazol-3-one SMILES: C1C(=NN(C1=O)C2=CC=CC=C2)N