accessibility menu, dialog, popup

UserName

CAS RN 4146-73-0

OFFFOOFFFOPb

CAS RN 4146-73-0

IUPAC Name: λ²-lead(2+) ditrifluoroacetate
Synonyms: λ²-lead(2+) ditrifluoroacetate
Molecular Weight (g/mol): 433.2
Molecular Formula: C4F6O4Pb
InChi Key: XVULNTYAIWEMSW-UHFFFAOYSA-L
SMILES: [Pb++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results