accessibility menu, dialog, popup

UserName

ClOClClCl

CAS RN 4136-95-2

IUPAC Name: 2,4,6-trichlorobenzoyl chloride
Synonyms: 2,4,6-trichlorobenzoyl chloride
Molecular Weight (g/mol): 243.89
Molecular Formula: C7H2Cl4O
InChi Key: OZGSEIVTQLXWRO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)